Learn More
Chemglass Life Sciences Bottle, Washing, Self-Venting, Safety-Labeled, Wide-Mouth, LDPE, 500 mL (Acetone) - Pack of 6
SDP
Catalog No. NC2959217
Encompass_Preferred
This 500 mL LDPE wash bottle is designed for dispensing and rinsing laboratory solvents. It features a wide mouth for easier filling, a one-piece closure and stem to reduce leakage, and a non-metallic self-venting design to minimize solvent leakage risk.
  • Capacity: 500 mL.
  • Material: LDPE bottle; polypropylene (PP) closure and stem.
  • Venting: self-venting with non-metallic vent.
  • Labeling: safety labeled for acetone with printed CAS, NFPA codes, and hazard information.
  • Packaging: pack of 6.
  • Design features: wide mouth for easy filling; angled neck and cut-back tip option; leak-resistant one-piece closure and stem.
Catalog No. NC2959217 Supplier Chemglass Life Sciences Supplier No. CG1131V001
This 500 mL LDPE wash bottle is designed for dispensing and rinsing laboratory solvents. It features a wide mouth for easier filling, a one-piece closure and stem to reduce leakage, and a non-metallic self-venting design to minimize solvent leakage risk.
  • Capacity: 500 mL.
  • Material: LDPE bottle; polypropylene (PP) closure and stem.
  • Venting: self-venting with non-metallic vent.
  • Labeling: safety labeled for acetone with printed CAS, NFPA codes, and hazard information.
  • Packaging: pack of 6.
  • Design features: wide mouth for easy filling; angled neck and cut-back tip option; leak-resistant one-piece closure and stem.