Learn More
ACROBiosystems ACROBiosystems Mouse BTLA Protein, His Tag
Supplier:  ACROBiosystems BTAM52E250UG
Encompass_Preferred
Recombinant Protein;50UG;Mouse BTLA, His Tag (BTA-M52E2) is expressed from human 293 cells (HEK293). It contains AA Glu 30 - Gly 176 (Accession # Q32MV9).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.6 kDa. The protein migrates as 30-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse BTLA, His Tag (Cat. No. BTA-M52E2) at 5 μg/mL (100 μL/well) can bind Mouse B7-H4, Fc Tag (Cat. No. B74-M5257) with a linear range of 0.05-0.8 μg/mL (QC tested).
Catalog No. 50-195-7293