Learn More
AdipoGen anti-BTLA (human), mAb (6F4),Monoclonal Antibody.
SDP
Supplier:  AdipoGen AG20B0049C100
Encompass_Preferred
anti-BTLA (human), mAb (6F4), AdipoGen Life Sciences. Monoclonal Antibody. Recognizes human BTLA. Isotype: Rat IgG1. Clone: 6F4. Applications: ELISA, FACS, FUNC (Blocking). Liquid. In PBS containing 10% glycerol.
Catalog No. 50-114-5717