Learn More
Sino Biological Recombinant Human BTLA Protein (ECD, mFc Tag)
Supplier:  Sino Biological 11895H38H200
Encompass_Preferred
A DNA sequence encoding the human BTLA (AAI07092.1) (Met1-Thr134) was expressed with the Fc region of mouse IgG2a at the C-terminus.
Catalog No. 50-211-5749