Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human BTLA (31-150) Protein, Fc Tag, Avi Tag (Avitag™);HEK293;200UG;BTA,BTLA,CD272
Catalog No. 501657693
Encompass_Preferred
Recombinant Protein;Biotinylated Human BTLA (31-150) Protein, Fc Tag, Avi Tag (Avitag™);200UG;Biotinylated Human BTLA (31-150), Fc Tag, Avi Tag (BTA-H82F3) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Ser 150 (Accession # AAP44003.1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 42.5 kDa. The protein migrates as 55-66 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human HVEM, His Tag (Cat. No. HVM-H52E9) at 5 μg/mL (100 μL/well) can bind Biotinylated Human BTLA (31-150), Fc Tag, Avi Tag (Cat. No. BTA-H82F3) with a linear range of 0.02-0.625 μg/mL (QC tested).
Catalog No. 50-165-7693 Supplier ACROBiosystems Supplier No. BTAH82F3200UG
Recombinant Protein;Biotinylated Human BTLA (31-150) Protein, Fc Tag, Avi Tag (Avitag™);200UG;Biotinylated Human BTLA (31-150), Fc Tag, Avi Tag (BTA-H82F3) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Ser 150 (Accession # AAP44003.1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 42.5 kDa. The protein migrates as 55-66 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human HVEM, His Tag (Cat. No. HVM-H52E9) at 5 μg/mL (100 μL/well) can bind Biotinylated Human BTLA (31-150), Fc Tag, Avi Tag (Cat. No. BTA-H82F3) with a linear range of 0.02-0.625 μg/mL (QC tested).