Learn More
Medchemexpress LLC BTMPS | 52829-07-9 | 98.0% | 480.73 | 100 G
SDP
Catalog No. 5000454285
Encompass_Preferred
BTMPS is a nicotine receptor (nAChR) antagonist that can lessen the negative effects of morphine on rats. It is for research use only and not sold to patients.
  • Acts as a nicotine receptor (nAChR) antagonist
  • Can lessen the negative effects of morphine on rats
  • For research use only
Catalog No. 50-004-54285 Supplier Medchemexpress LLC Supplier No. HYW013113100G
BTMPS is a nicotine receptor (nAChR) antagonist that can lessen the negative effects of morphine on rats. It is for research use only and not sold to patients.
  • Acts as a nicotine receptor (nAChR) antagonist
  • Can lessen the negative effects of morphine on rats
  • For research use only