Learn More
Carnabio Usa Inc BTN - ACTR2B(ACVR2B) / 10UG
Supplier:  Carnabio Usa Inc 0943220N10
Encompass
BTN-ACTR2B(ACVR2B), Cytoplasmic domain, Wild type, Amino Acid 161-512(end), NP_001097.2, 43 kDa, Insect (sf21); 10microg
Catalog No. 50-155-993