Learn More
Carnabio Usa Inc BTN - AKT1 / 10UG
Catalog No. 507541069
Encompass
BTN-AKT1, Catalytic domain, Wild type, Amino Acid 104-480(end), NP_005154.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-1069 Supplier Carnabio Usa Inc Supplier No. 0140120N10
BTN-AKT1, Catalytic domain, Wild type, Amino Acid 104-480(end), NP_005154.1, Expressed in Insect (sf21); 10microg