Learn More
Carnabio Usa Inc BTN - EGFR[T790M/L858RÙ/ 10UG
Catalog No. 50155995
Encompass
BTN-EGFR[T790M/L858R], Cytoplasmic domain, Wild type, Amino Acid 669-1210(end), NP_005219.2, 66 kDa, Insect (sf21); 10microg
Catalog No. 50-155-995 Supplier Carnabio Usa Inc Supplier No. 0841020N10
BTN-EGFR[T790M/L858R], Cytoplasmic domain, Wild type, Amino Acid 669-1210(end), NP_005219.2, 66 kDa, Insect (sf21); 10microg