Learn More
Carnabio Usa Inc BTN - FMS(CSF1R) / 10UG
Supplier:  Carnabio Usa Inc 0845520N10
Encompass
BTN-FMS(CSF1R), Cytoplasmic domain, Wild type, Amino Acid 538-972(end), NP_005202.2, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-1076