Learn More
Carnabio Usa Inc BTN - IKKA(CHUK) / 10UG
Catalog No. 507541079
Encompass
BTN-IKK(CHUK), Full-length, Wild type, Amino Acid 1-745(end), NP_001269.3, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-1079 Supplier Carnabio Usa Inc Supplier No. 0541220N10
BTN-IKK(CHUK), Full-length, Wild type, Amino Acid 1-745(end), NP_001269.3, Expressed in Insect (sf21); 10microg