Learn More
Carnabio Usa Inc BTN - IRAK4 / 10UG
Catalog No. 50156000
Encompass
BTN-IRAK4, Full-length, Wild type, Amino Acid 1-460(end), NP_057207.2, 55 kDa, Insect (sf21); 10microg
Catalog No. 50-156-000 Supplier Carnabio Usa Inc Supplier No. 0944520N10
BTN-IRAK4, Full-length, Wild type, Amino Acid 1-460(end), NP_057207.2, 55 kDa, Insect (sf21); 10microg