Learn More
Carnabio Usa Inc BTN - RET / 10UG
Catalog No. 50156004
Encompass
BTN-RET, Cytoplasmic domain, Wild type, Amino Acid 658-1114(end), NP_066124.1, 56 kDa, Insect (sf21); 10microg
Catalog No. 50-156-004 Supplier Carnabio Usa Inc Supplier No. 0845920N10
BTN-RET, Cytoplasmic domain, Wild type, Amino Acid 658-1114(end), NP_066124.1, 56 kDa, Insect (sf21); 10microg