Learn More
ACROBiosystems ACROBiosystems Human BTN1A1 / Butyrophilin Protein, Fc Tag
Supplier:  ACROBiosystems BT1H52531MG
Encompass_Preferred
Recombinant Protein;1MG;Human BTN1A1, Fc Tag (BT1-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 242 (Accession # AAH96312).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50.6 kDa. As a result of glycosylation, the protein migrates as 55-66 kDa under reducing (R) condition, and 116-130 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-195-7509