Learn More
ACROBiosystems ACROBiosystems Human BTN1A1 / Butyrophilin Protein, His Tag
Supplier:  ACROBiosystems BT1H52221MG
Encompass_Preferred
Recombinant Protein;1MG;Human BTN1A1, His Tag (BT1-H5222) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 242 (Accession # AAH96312).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.8 kDa. The protein migrates as 35-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7511