Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human BTN1A1 / Butyrophilin Protein, His Tag & Avi Tag (Avitag™);HEK293;25UG;BT1,BTN1A1,Butyrophilin,BTN,BT
Supplier: ACROBiosystems BT1H82E625UG
Recombinant Protein;Biotinylated Human BTN1A1 / Butyrophilin Protein, His Tag & Avi Tag (Avitag™);25UG;Biotinylated Human BTN1A1, His Tag (BT1-H82E6) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 242 (Accession # Q13410-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 27.6 kDa. The protein migrates as 36-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.