Learn More
Aviva Systems Biology Corp BTN2A1 Antibody - N-terminal region (ARP46817_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46817_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BTN2A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human BTN2A1. Alternate names: BK14H9.1, BT2.1, BTF1, DJ3E1.1, FLJ36567ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59412