Learn More
Genscript Corporation BTN3A1/CD277Biotin His Av
Catalog No. 5000058691
Encompass
The three butyrophilin BTN3A molecules BTN3A1 BTN3A2 and BTN3A3 are members of the B7/butyrophilin-like group of Ig superfamily receptors which modulate the function of T cells BTN3A1 controls activation of human V 9/V 2 T cells by direct or indirect presentation of self and nonself phosphoantigens (pAg)
Catalog No. 50-000-58691 Supplier Genscript Corporation Supplier No. Z04359500
The three butyrophilin BTN3A molecules BTN3A1 BTN3A2 and BTN3A3 are members of the B7/butyrophilin-like group of Ig superfamily receptors which modulate the function of T cells BTN3A1 controls activation of human V 9/V 2 T cells by direct or indirect presentation of self and nonself phosphoantigens (pAg)