Learn More
ACROBiosystems ACROBiosystems Human BTN3A1 / CD277 Protein, Fc Tag
Supplier:  ACROBiosystems BT1H5255100UG
Encompass_Preferred
Recombinant Protein;100UG;Human BTN3A1, Fc Tag (BT1-H5255) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Gly 254 (Accession # AAI21801).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50.8 kDa. As a result of glycosylation, the protein migrates as 58-65 kDa under reducing (R) condition, and 120 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-195-7507