Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human BTN3A1 / CD277 Protein, Fc Tag, Avi Tag (Avitag™);HEK293;25UG;BT1,BTN3A1,CD277,BTF5,Butyrophilin
Catalog No. 501658084
Encompass_Preferred
Recombinant Protein;Biotinylated Human BTN3A1 / CD277 Protein, Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Human BTN3A1, Fc Tag, Avi Tag (BT1-H82F7) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Gly 254 (Accession # O00481-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 52.9 kDa. The protein migrates as 54 kDa and 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8084 Supplier ACROBiosystems Supplier No. BT1H82F725UG
Recombinant Protein;Biotinylated Human BTN3A1 / CD277 Protein, Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Human BTN3A1, Fc Tag, Avi Tag (BT1-H82F7) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Gly 254 (Accession # O00481-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 52.9 kDa. The protein migrates as 54 kDa and 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.