Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human BTN3A2 Protein, His Tag, Avi Tag (Avitag™);HEK293;200UG;BT2,BT3.2,BT3.3,BTF3,BTF4,BTN3A2,FLJ40011
Supplier:  ACROBiosystems BT2H82E7200UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human BTN3A2 Protein, His Tag, Avi Tag (Avitag™);200UG;Biotinylated Human BTN3A2, His Tag, Avi Tag (BT2-H82E7) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Trp 248 (Accession # P78410-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 27.3 kDa. The protein migrates as 30-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7677