Learn More
ACROBiosystems ACROBiosystems Human BTN3A3 / BTF3 Protein, His Tag
Supplier:  ACROBiosystems BT3H52H0100UG
Encompass_Preferred
Recombinant Protein;100UG;Human BTN3A3, His Tag (BT3-H52H0) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Trp 248 (Accession # AAH15815).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.5 kDa. The protein migrates as 28 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7506