Learn More
ACROBiosystems Recombinant Protein;Human BTN3A3 / BTF3 Protein;HEK293;1MG;BT3,BTN3A3,BTF3,BTN3A-3,BTF-3
Catalog No. 501659001
Encompass_Preferred
Recombinant Protein;Human BTN3A3 / BTF3 Protein;1MG;Human BTN3A3, His Tag (BT3-H52H0) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Trp 248 (Accession # AAH15815).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.5 kDa. The protein migrates as 28 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-9001 Supplier ACROBiosystems Supplier No. BT3H52H01MG
Recombinant Protein;Human BTN3A3 / BTF3 Protein;1MG;Human BTN3A3, His Tag (BT3-H52H0) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Trp 248 (Accession # AAH15815).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.5 kDa. The protein migrates as 28 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.