Learn More
Aviva Systems Biology Corp BTNL3 Antibody - N-terminal region (ARP46769_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46769_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BTNL3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human BTNL3. Alternate names: BTNLRApplicationWB. Species reactivity: Cow, Horse, Human, Pig
Catalog No. 50-014-59395