Learn More
Aviva Systems Biology Corp BTNL9 Antibody - N-terminal region (ARP53034_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53034_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BTNL9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human BTNL9. Alternate names: BTN3, VDLS1900ApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-014-60899