Learn More
BTNX Inc BTNX 3-Drug Test Panel 25/Kit XYL10 FYL10 NTZ500
Catalog No. NC3957414
Encompass_Preferred
The Rapid Response Multi-Drug Test Panel is a rapid test for the simultaneous detection of multiple drugs in human urine. The test can detect for Xylazine (XYL) at a cut-off level of 10 ng/mL, Fentanyl (FYL) at a cut-off level of 10 ng/mL, and Nitazene (NTZ) at a cut-off level of 500 ng/mL. This test provides only a qualitative preliminary analytical test result only.
Catalog No. NC3957414 Supplier BTNX Inc Supplier No. D3.381P2925
The Rapid Response Multi-Drug Test Panel is a rapid test for the simultaneous detection of multiple drugs in human urine. The test can detect for Xylazine (XYL) at a cut-off level of 10 ng/mL, Fentanyl (FYL) at a cut-off level of 10 ng/mL, and Nitazene (NTZ) at a cut-off level of 500 ng/mL. This test provides only a qualitative preliminary analytical test result only.