Learn More
BTNX Inc BTNX 3-in-1 Spiked Drink Pen 5/Kit BZO600ng/mL KET2000ng/mL GHB1mg/mL
Catalog No. NC3957516
Encompass_Preferred
The Rapid Response 3-in-1 Spiked Drink Pen detects Benzodiazepines at 600 ng/mL, Ketamine at 2000 ng/mL and GHB at 1 mg/mL in beverages. Quantity: 5/kit. Time to result: 3 minutes for GHB, 5 minutes for BZO/KET.
Catalog No. NC3957516 Supplier BTNX Inc Supplier No. D3.398DH5
The Rapid Response 3-in-1 Spiked Drink Pen detects Benzodiazepines at 600 ng/mL, Ketamine at 2000 ng/mL and GHB at 1 mg/mL in beverages. Quantity: 5/kit. Time to result: 3 minutes for GHB, 5 minutes for BZO/KET.