Learn More
BTNX Inc BTNX Benzodiazepine Test Strips (Liquid/Powder) 10/Kit Cutoff 300 ng/mL
Catalog No. NC3957522
Encompass_Preferred
The Rapid Response™ Benzodiazepine Test Strip (Liquid / Powder) is a rapid visual immunoassay for the qualitative, presumptive detection of benzodiazepine in suspicious substances (liquids or powders) at the cut-off concentration of 300 ng/mL. Easy to use with detailed instructions for testing both liquid and powder samples. Quantity: 10/Kit. Time to result: 5 minutes.
Catalog No. NC3957522 Supplier BTNX Inc Supplier No. BZO18S310
The Rapid Response™ Benzodiazepine Test Strip (Liquid / Powder) is a rapid visual immunoassay for the qualitative, presumptive detection of benzodiazepine in suspicious substances (liquids or powders) at the cut-off concentration of 300 ng/mL. Easy to use with detailed instructions for testing both liquid and powder samples. Quantity: 10/Kit. Time to result: 5 minutes.