Learn More
BTNX Inc BTNX CLIA-Waived 10-Drug Test Panel 25/Kit COC300 AMP1000 MET1000 THC50 MTD300 OPI2000 PCP25 BAR300 BZO300 TCA1000
Catalog No. NC3957418
Encompass_Preferred
The Rapid Response 10 Parameter Multi-Drug Test Panel is a one step, qualitative test for the simultaneous detection of COC300 AMP1000 MET1000 THC50 MTD300 OPI2000 PCP25 BAR300 BZO300 TCA1000 in human urine. In just 5 minutes, test results can be observed through the visual interpretation of coloured lines. Quantity: 25/kit. CLIA-Waived.
Catalog No. NC3957418 Supplier BTNX Inc Supplier No. D10.11P2925
The Rapid Response 10 Parameter Multi-Drug Test Panel is a one step, qualitative test for the simultaneous detection of COC300 AMP1000 MET1000 THC50 MTD300 OPI2000 PCP25 BAR300 BZO300 TCA1000 in human urine. In just 5 minutes, test results can be observed through the visual interpretation of coloured lines. Quantity: 25/kit. CLIA-Waived.