Learn More
BTNX Inc BTNX CLIA-Waived 12-Drug Test Panel 25/Kit COC300 AMP1000 MET1000 THC50 MTD300 MDMA500 MOR300 OXY100 PPX300 BAR300 BZO300 BUP10
Catalog No. NC3957419
Encompass_Preferred
The Rapid Response 12 Parameter Multi-Drug Test Panel is a one step, qualitative test for the simultaneous detection of COC300 AMP1000 MET1000 THC50 MTD300 MDMA500 MOR300 OXY100 PPX300 BAR300 BZO300 BUP10 in human urine. In just 5 minutes, test results can be observed through the visual interpretation of coloured lines. Quantity: 25/kit. CLIA-Waived.
Catalog No. NC3957419 Supplier BTNX Inc Supplier No. D12.11P2925
The Rapid Response 12 Parameter Multi-Drug Test Panel is a one step, qualitative test for the simultaneous detection of COC300 AMP1000 MET1000 THC50 MTD300 MDMA500 MOR300 OXY100 PPX300 BAR300 BZO300 BUP10 in human urine. In just 5 minutes, test results can be observed through the visual interpretation of coloured lines. Quantity: 25/kit. CLIA-Waived.