Learn More
BTNX Inc BTNX CLIA-Waived 5-Drug Test Panel 25/Kit COC300 AMP1000 MET1000 THC50 OPI2000
Catalog No. NC3957415
Encompass_Preferred
The Rapid Response 5 Parameter Multi-Drug Test Panel is a one step, qualitative test for the simultaneous detection of COC300 AMP1000 MET1000 THC50 OPI2000 in human urine. In just 5 minutes, test results can be observed through the visual interpretation of coloured lines. Quantity: 25/kit. CLIA-Waived.
Catalog No. NC3957415 Supplier BTNX Inc Supplier No. D5.11P2925
The Rapid Response 5 Parameter Multi-Drug Test Panel is a one step, qualitative test for the simultaneous detection of COC300 AMP1000 MET1000 THC50 OPI2000 in human urine. In just 5 minutes, test results can be observed through the visual interpretation of coloured lines. Quantity: 25/kit. CLIA-Waived.