Learn More
BTNX Inc BTNX CLIA-Waived Drug Test Cup 13-Panel with Fentanyl (Flat Cup Format) OPI300 MDMA500 OXY100 PCP25 MTD300 BAR300 BZO300 COC150 AMP500 MET500 THC50 BUP10 FYL1
Catalog No. NC3957428
Encompass_Preferred
The Rapid Response™ 13 Parameter Multi-drug One Step Flat Cup is a CLIA Waived, one-step rapid test that detects OPI300 MDMA500 OXY100 PCP25 MTD300 BAR300 BZO300 COC150 AMP500 MET500 THC50 BUP10 FYL1 in urine and is now enhanced with the addition of a fentanyl test strip. With the rising prevalence of fentanyl in the drug supply, incorporating fentanyl testing into our CLIA Waived test cups is a critical feature, providing you with accurate and rapid results that meet your testing needs. Quantity: 25/kit.The flat cup design is compatable with the Rapid Response Workflow solutions: The RapidReader App and Lateral Flow Scanner. Designed to help clinics like yours streamline their drug screening process with AI-powered drug screen recording and data management.
Catalog No. NC3957428 Supplier BTNX Inc Supplier No. D13.51VF
The Rapid Response™ 13 Parameter Multi-drug One Step Flat Cup is a CLIA Waived, one-step rapid test that detects OPI300 MDMA500 OXY100 PCP25 MTD300 BAR300 BZO300 COC150 AMP500 MET500 THC50 BUP10 FYL1 in urine and is now enhanced with the addition of a fentanyl test strip. With the rising prevalence of fentanyl in the drug supply, incorporating fentanyl testing into our CLIA Waived test cups is a critical feature, providing you with accurate and rapid results that meet your testing needs. Quantity: 25/kit.The flat cup design is compatable with the Rapid Response Workflow solutions: The RapidReader App and Lateral Flow Scanner. Designed to help clinics like yours streamline their drug screening process with AI-powered drug screen recording and data management.