Learn More
BTNX Inc BTNX CLIA-Waived Drug Test Cup 14-Panel (Flat Cup Format) AMP1000 BAR300 BUP10 BZO300 COC300 MET1000 MDMA500 OPI2000 MTD300 OXY100 PCP25 THC50 PPX300 TCA1000
Catalog No. NC3957442
Encompass_Preferred
The Rapid Response 14 Parameter Multi-Drug One Step Cup (Flat) is an easy-to-use, one-step solution for drug screening that can simultaneously detect multiple drugs in urine samples. This format is a great alternative to the industry's complicated testing processes and is available with and without adulterant tests. It has a longer detection window than saliva tests and provides results in 5 minutes. Quantity: 25/kit. CLIA-Waived.Rapid Response 14 Parameter Multi-Drug One Step Cup (Flat) is compatible with our RapidReader App, allowing users to upload results to their ATS/EMR/EHR in seconds without the paperwork.
Catalog No. NC3957442 Supplier BTNX Inc Supplier No. D14.51VF
The Rapid Response 14 Parameter Multi-Drug One Step Cup (Flat) is an easy-to-use, one-step solution for drug screening that can simultaneously detect multiple drugs in urine samples. This format is a great alternative to the industry's complicated testing processes and is available with and without adulterant tests. It has a longer detection window than saliva tests and provides results in 5 minutes. Quantity: 25/kit. CLIA-Waived.Rapid Response 14 Parameter Multi-Drug One Step Cup (Flat) is compatible with our RapidReader App, allowing users to upload results to their ATS/EMR/EHR in seconds without the paperwork.