Learn More
BTNX Inc BTNX CLIA-Waived Fecal Immunochemical Test (FIT) 36 Tests/Kit
Catalog No. NC3957481
Encompass_Preferred
The Rapid Response Fecal Immunochemical Test is a rapid, qualitative test for the determination of human occult blood in feces, to aid in the early detection of gastrointestinal problems such as colorectal cancer. The test may be used during routine physical examinations, in hospitals to monitor patient bleeding, or as a screening tool for colorectal cancer or gastrointestinal bleeding from any source. Unlike guaiac assays, the accuracy of this test is not affected by the diet of the patients. It is intended for in vitro diagnostic use only. CLIA-Waived. Time to result: 5 minutes.Kit contains 36 tests and 36 buffer tubes.
Catalog No. NC3957481 Supplier BTNX Inc Supplier No. FOB9C36
The Rapid Response Fecal Immunochemical Test is a rapid, qualitative test for the determination of human occult blood in feces, to aid in the early detection of gastrointestinal problems such as colorectal cancer. The test may be used during routine physical examinations, in hospitals to monitor patient bleeding, or as a screening tool for colorectal cancer or gastrointestinal bleeding from any source. Unlike guaiac assays, the accuracy of this test is not affected by the diet of the patients. It is intended for in vitro diagnostic use only. CLIA-Waived. Time to result: 5 minutes.Kit contains 36 tests and 36 buffer tubes.