Learn More
BTNX Inc BTNX CLIA-Waived Midstream Pregnancy Test 1 Test/Kit hCG 20 mIU
Catalog No. NC3957507
Encompass_Preferred
The Rapid Response Midstream Pregnancy Test is a rapid, qualitative immunoassay for the determination of human chorionic gonadotropin (hCG) in urine specimen at 20 mIU/mL or above. This kit is intended for use as an aid in the early detection of pregnancy. Sensitivity: 20 mIU/mL. Quantity: 1 test/kit. CLIA-Waived.
Catalog No. NC3957507 Supplier BTNX Inc Supplier No. HCG1M1
The Rapid Response Midstream Pregnancy Test is a rapid, qualitative immunoassay for the determination of human chorionic gonadotropin (hCG) in urine specimen at 20 mIU/mL or above. This kit is intended for use as an aid in the early detection of pregnancy. Sensitivity: 20 mIU/mL. Quantity: 1 test/kit. CLIA-Waived.