Learn More
BTNX Inc BTNX CLIA-Waived Pregnancy Strips 50 Strips/Kit hCG 20 mIU
Catalog No. NC3957509
Encompass_Preferred
The Rapid Response™ Pregnancy Test Strip is a rapid visual immunoassay for the qualitative, presumptive detection of human chorionic gonadotropin in human urine specimens. This kit is intended for use as an aid in the early detection of pregnancy. Sensitivity: 20 mIU/mL. Quantity: 50/kit. CLIA-Waived.
Catalog No. NC3957509 Supplier BTNX Inc Supplier No. HCG1S50
The Rapid Response™ Pregnancy Test Strip is a rapid visual immunoassay for the qualitative, presumptive detection of human chorionic gonadotropin in human urine specimens. This kit is intended for use as an aid in the early detection of pregnancy. Sensitivity: 20 mIU/mL. Quantity: 50/kit. CLIA-Waived.