Learn More
BTNX Inc BTNX CLIA-Waived Urine Analyzer (Smart with Barcode Reader) 1 Unit
Supplier:  BTNX Inc UAU121S
Encompass_Preferred
Streamline your clinic’s testing with the Rapid Response™ U121S Urine Analyzer – an efficient and accurate tool for interpreting Rapid Response™ Urinalysis Reagent Strips results. With the ability to swiftly read, store, and print results in under two minutes, the U121S Urine Analyzer improves workflow efficiency while reducing the chance of human error. Rapid Response™ U121S Urine Analyzer is intended to be used in conjunction with Rapid Response™ Urinalysis Reagent Strips. Together they can be used to aid in the general evaluation of health, the diagnosis and monitoring of metabolic, systemic, or endocrine disorders affecting kidney function, and detecting diseases of the urinary tract. For professional in vitro diagnostic use only. CLIA-Waived.Ideal for clinics with a high throughput, the Rapid Response™ U121S, conveniently includes a barcode reader offering quick canister code recognition and Patient ID entry.
Catalog No. NC3957498