Learn More
BTNX Inc BTNX Fecal Immunochemical Test (FIT) Collection Buffer Tubes 30 Buffers/Kit
Catalog No. NC3957488
Encompass_Preferred
The Rapid Response™ Fecal Immunochemical Test (FIT) Buffer/Collection Tubes are to be used with corresponding Rapid Response™ FIT - Fecal Immunochemical Test Kits for sample collection and preparation. The tubes are specially designed to prevent overloading of the sample tube, effectively reducing invalid results and are unique in the marketplace. Quantity: 30/kit. CLIA-Waived.
Catalog No. NC3957488 Supplier BTNX Inc Supplier No. FOB9V30
The Rapid Response™ Fecal Immunochemical Test (FIT) Buffer/Collection Tubes are to be used with corresponding Rapid Response™ FIT - Fecal Immunochemical Test Kits for sample collection and preparation. The tubes are specially designed to prevent overloading of the sample tube, effectively reducing invalid results and are unique in the marketplace. Quantity: 30/kit. CLIA-Waived.