Learn More
BTNX Inc BTNX Fecal Immunochemical Test (FIT) Mailers 50 Mailers/Box
Catalog No. NC3957490
Encompass_Preferred
The Rapid Response Fecal Immunochemical Test (FIT) Mailer is a paper kit designed to be given to patients and used in conjunction with the Rapid Response FIT Test for at-home collection of spontaneously passed stool (SPS) specimens. This easy collection method increases patient compliance and ensures appropriate sample collection. Quantity: 50/kit.
Catalog No. NC3957490 Supplier BTNX Inc Supplier No. FOB0M
The Rapid Response Fecal Immunochemical Test (FIT) Mailer is a paper kit designed to be given to patients and used in conjunction with the Rapid Response FIT Test for at-home collection of spontaneously passed stool (SPS) specimens. This easy collection method increases patient compliance and ensures appropriate sample collection. Quantity: 50/kit.