Learn More
BTNX Inc BTNX Liquid Diptube Urine Control Kit 2x12mL Vials
Catalog No. NC3957506
Encompass_Preferred
The Rapid Response Liquid Diptube Urine Controls contains one Level 1 Control Tube (12 mL) and one Level 2 Control Tube (12 mL) and are intended for use in validating the visual and analyzer reading of urinalysis, to ensure the consistent performance of Rapid Response Urinalysis Reagent Strips and Rapid Response U120 Urine Analyzers. They are intended for in vitro diagnostic use only. For visual reading, test results are interpreted by direct comparison of the color blocks printed on the canister label. For Analyzer reading, refer to the Instruction Manual for details.
Catalog No. NC3957506 Supplier BTNX Inc Supplier No. AUCT1V1
The Rapid Response Liquid Diptube Urine Controls contains one Level 1 Control Tube (12 mL) and one Level 2 Control Tube (12 mL) and are intended for use in validating the visual and analyzer reading of urinalysis, to ensure the consistent performance of Rapid Response Urinalysis Reagent Strips and Rapid Response U120 Urine Analyzers. They are intended for in vitro diagnostic use only. For visual reading, test results are interpreted by direct comparison of the color blocks printed on the canister label. For Analyzer reading, refer to the Instruction Manual for details.