Learn More
BTNX Inc BTNX Saliva Drug Test Cup 10-Panel (Flat Cup Format) COC20 OPI40 AMP50 MET50 BAR50 BZO10 MDMA50 MTD30 PCP10 OXY20
Catalog No. NC3957461
Encompass_Preferred
The Rapid Response™ 10 Parameter Multi-Drug Saliva Test Cup detects THC40, COC20, OPI40, AMP50, MET50, BAR50, BZO10, MDMA50, MTD30, PCP10 in oral fluid. Includes saturation indicator. Compatible with The RapidReader App, allowing users to upload results to their ATS/EMR/EHR in seconds without the paperwork. Quantity: 25/kit. Time to result: 10 minutes.
Catalog No. NC3957461 Supplier BTNX Inc Supplier No. D10.32.12FC1
The Rapid Response™ 10 Parameter Multi-Drug Saliva Test Cup detects THC40, COC20, OPI40, AMP50, MET50, BAR50, BZO10, MDMA50, MTD30, PCP10 in oral fluid. Includes saturation indicator. Compatible with The RapidReader App, allowing users to upload results to their ATS/EMR/EHR in seconds without the paperwork. Quantity: 25/kit. Time to result: 10 minutes.