Learn More
BTNX Inc BTNX Saliva Drug Test Cup 5-Panel (Flat Cup Format) COC20 OPI40 AMP50 MET50 OXY20
Catalog No. NC3957453
Encompass_Preferred
The Rapid Response™ 5 Parameter Multi-Drug Saliva Test Cup detects COC20, OPI40, AMP50, MET50, OXY20 in oral fluid. Includes saturation indicator. Compatible with The RapidReader App, allowing users to upload results to their ATS/EMR/EHR in seconds without the paperwork. Quantity: 25/kit. Time to Result: 10 minutes.
Catalog No. NC3957453 Supplier BTNX Inc Supplier No. D5.75.12FC1
The Rapid Response™ 5 Parameter Multi-Drug Saliva Test Cup detects COC20, OPI40, AMP50, MET50, OXY20 in oral fluid. Includes saturation indicator. Compatible with The RapidReader App, allowing users to upload results to their ATS/EMR/EHR in seconds without the paperwork. Quantity: 25/kit. Time to Result: 10 minutes.