Learn More
BTNX Inc BTNX Saliva Drug Test Cup 6-Panel (Flat Cup Format) COC20 MET50 PCP10 THC40 OPI40 AMP50
Catalog No. NC3957455
Encompass_Preferred
The Rapid Response 6 Parameter Multi-Drug Saliva Test Cup is a rapid, qualitative screening test for the simultaneous detection of COC20 MET50 PCP10 THC40 OPI40 AMP50 in human oral fluid. An indicator strip is included in the indicator window of the collection swab to signal saturation. Quantity: 25/kit. Time to result: 10 minutes.Rapid Response Multi-Drug Saliva Test Cup is compatible with The RapidReader App, allowing users to upload results to their ATS/EMR/EHR in seconds without the paperwork.
Catalog No. NC3957455 Supplier BTNX Inc Supplier No. D6.3.12FC1
The Rapid Response 6 Parameter Multi-Drug Saliva Test Cup is a rapid, qualitative screening test for the simultaneous detection of COC20 MET50 PCP10 THC40 OPI40 AMP50 in human oral fluid. An indicator strip is included in the indicator window of the collection swab to signal saturation. Quantity: 25/kit. Time to result: 10 minutes.Rapid Response Multi-Drug Saliva Test Cup is compatible with The RapidReader App, allowing users to upload results to their ATS/EMR/EHR in seconds without the paperwork.