Learn More
BTNX Inc BTNX Saliva Drug Test Cup 9-Panel (Flat Cup Format) COC20 OPI40 AMP50 MET50 BAR50 BZO10 MDMA50 MTD30 PCP10
Catalog No. NC3957457
Encompass_Preferred
The Rapid Response™ 9 Parameter Multi-Drug Saliva Test Cup detects COC20, OPI40, AMP50, MET50, BAR50, BZO10, MDMA50, MTD30, PCP10 in oral fluid. Includes saturation indicator. Compatible with The RapidReader App, allowing users to upload results to their ATS/EMR/EHR in seconds without the paperwork. Quantity: 25/kit. Time to result: 10 minutes.
Catalog No. NC3957457 Supplier BTNX Inc Supplier No. D9.172FC1
The Rapid Response™ 9 Parameter Multi-Drug Saliva Test Cup detects COC20, OPI40, AMP50, MET50, BAR50, BZO10, MDMA50, MTD30, PCP10 in oral fluid. Includes saturation indicator. Compatible with The RapidReader App, allowing users to upload results to their ATS/EMR/EHR in seconds without the paperwork. Quantity: 25/kit. Time to result: 10 minutes.