Learn More
BTNX Inc BTNX Saliva Drug Test Stick 10-Panel 25/Kit COC20 MET50 THC40 BZO10 AMP50 OPI40 MDMA50 MTD30 BAR50 PCP10
Catalog No. NC3957479
Encompass_Preferred
The Rapid Response 10 Parameter Oral Fluid Lollipop Drug Test detects COC20, MET50, THC40, BZO10, AMP50, OPI40, MDMA50, MTD30, BAR50, PCP10 in saliva. Results in 10 minutes. Compatible with The RapidReader App, allowing users to upload results to their ATS/EMR/EHR in seconds without the paperwork. Quantity: 25/kit.
Catalog No. NC3957479 Supplier BTNX Inc Supplier No. D10.3.12L25
The Rapid Response 10 Parameter Oral Fluid Lollipop Drug Test detects COC20, MET50, THC40, BZO10, AMP50, OPI40, MDMA50, MTD30, BAR50, PCP10 in saliva. Results in 10 minutes. Compatible with The RapidReader App, allowing users to upload results to their ATS/EMR/EHR in seconds without the paperwork. Quantity: 25/kit.