Learn More
BTNX Inc BTNX Saliva Drug Test Stick 5-Panel 25/Kit THC40 COC20 AMP50 MET50 OPI40
Catalog No. NC3957477
Encompass_Preferred
The Rapid Response 5 Parameter Oral Fluid Lollipop Drug Test detects THC40, COC20, AMP50, MET50, OPI40 in saliva. Results in 10 minutes. Compatible with The RapidReader App, allowing users to upload results to their ATS/EMR/EHR in seconds without the paperwork. Quantity: 25/kit.
Catalog No. NC3957477 Supplier BTNX Inc Supplier No. D5.1.12L25
The Rapid Response 5 Parameter Oral Fluid Lollipop Drug Test detects THC40, COC20, AMP50, MET50, OPI40 in saliva. Results in 10 minutes. Compatible with The RapidReader App, allowing users to upload results to their ATS/EMR/EHR in seconds without the paperwork. Quantity: 25/kit.