Learn More
BTNX Inc BTNX Single Alcohol (ETG) Test Panel 25/Kit Ethyl Glucuronide (ETG) Cutoff 500 ng/mL
Supplier:  BTNX Inc ETG1P925
Encompass_Preferred
Rapid Response Ethyl Glucuronide (ETG) Test Panel detects ETG at 500 ng/mL cut-off in urine. ETG provides longer detection window for recent alcohol use. Rapid qualitative results in 5 minutes. For forensic use only. Quantity: 25/kit.
Catalog No. NC3957421