Learn More
BTNX Inc BTNX Single Drug Test Strip 100/Kit Ketamine (KET) Cutoff 300 ng/mL
Catalog No. NC3957395
Encompass_Preferred
Rapid Response Single Drug Test Strip for Ketamine (KET) detection in urine. Rapid, qualitative, targeted screening solution. Cut-off: 300 ng/mL. Time to result: 5 minutes. Quantity: 100/kit.
Catalog No. NC3957395 Supplier BTNX Inc Supplier No. KET1S3100
Rapid Response Single Drug Test Strip for Ketamine (KET) detection in urine. Rapid, qualitative, targeted screening solution. Cut-off: 300 ng/mL. Time to result: 5 minutes. Quantity: 100/kit.