Learn More
BTNX Inc BTNX Single Drug Test Strip 100/Kit Morphine (MOP) Cutoff 100 ng/mL
Catalog No. NC3957397
Encompass_Preferred
Rapid Response Single Drug Test Strip for Morphine (MOP) detection in urine. Rapid, qualitative, targeted screening solution. Cut-off: 100 ng/mL. Time to result: 5 minutes. Quantity: 25/kit.
Catalog No. NC3957397 Supplier BTNX Inc Supplier No. MOP1S27100
Rapid Response Single Drug Test Strip for Morphine (MOP) detection in urine. Rapid, qualitative, targeted screening solution. Cut-off: 100 ng/mL. Time to result: 5 minutes. Quantity: 25/kit.