Learn More
BTNX Inc BTNX Urinalysis Reagent Test Strip 2-Parameter (Glu/Pro) 100 Strips/Bottle
Catalog No. NC3957492
Encompass_Preferred
The Rapid Response 2-Parameter Urinalysis Reagent Strips are qualitative and semi-qualitative dip-and-read test for the detection of Glucose and Protein in human urine, to aid in the general evaluation of health and in the diagnosis and monitoring of metabolic or systemic diseases. Quantity: 100/bottle. Time to result: 30-60 seconds.
Catalog No. NC3957492 Supplier BTNX Inc Supplier No. U2.11S100
The Rapid Response 2-Parameter Urinalysis Reagent Strips are qualitative and semi-qualitative dip-and-read test for the detection of Glucose and Protein in human urine, to aid in the general evaluation of health and in the diagnosis and monitoring of metabolic or systemic diseases. Quantity: 100/bottle. Time to result: 30-60 seconds.